Prilocaine hydrochloride 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Prilocaine HCl, 2-(Propylamino)-N-(o-tolyl)propanamide hydrochloride, 2-(Propylamino)-N-(o-tolyl)propionamide Hydrochloride

J60648.03 J60648.06 J60648.09
J60648.03EA 76.8 EUR
J60648.03 J60648.06 J60648.09
Prilocaine hydrochloride 98%
Prilocaine hydrochloride

Purity: 98% CAS No.: 1786-81-8 Molecular Formula: C13H20N2O.HCl Formula Weight: 256.77 Physical Form: Powder Appearance: White Melting Point: 167-168° MDL No.: MFCD00079279


Applications: A local anesthetic

Formel: C₁₃H₂₀N₂O
Molecular Weight: 256.77 g/mol
Schmelzpunkt: 167…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079279
CAS-Nummer: 1786-81-8
EINECS: 217-244-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR