4-Thiouridine ≥98%
Lieferant: Thermo Scientific
Synonyme:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-4-thioxo-3,4-dihydropyrimidin-2(1H)-one
J60679.MC
J60679.MCEA
452
EUR
J60679.MC
4-Thiouridine ≥98%
4-Thiouridine
Purity: 98+%
CAS No.: 13957-31-8
Molecular Formula: C9H12N2O5S
Formula Weight: 260.27
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline
Appearance: Pale yellow
MDL No.: MFCD00006538
Formel:
C₉H₁₂N₂O₅S Molecular Weight: 260,264 g/mol |
MDL:
MFCD00006538 CAS-Nummer: 13957-31-8 EINECS: 237-735-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...