Hygromycin B

Lieferant: Thermo Fisher Scientific
Gefahr

J60681.MC J60681.MD J60681.03
J60681.MCEA 57.1 EUR
J60681.MC J60681.MD J60681.03
Hygromycin B
Hygromycin B
Formel: C₂₀H₃₇N₃O₁₃
Lagertemperatur: Kühlschrank
MDL: MFCD06795479
CAS-Nummer: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I
Merck Index: 14,4852

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR