GBR 12909 dihydrochloride

Lieferant: Thermo Scientific

J60684.MA
J60684.MAEA 117 EUR
J60684.MA
GBR 12909 dihydrochloride
GBR 12909 dihydrochloride

Dopamine uptake inhibitor.


  • Soluble in DMSO to 50mM. Soluble to 5mM in water with warming.


White powder

Formel: C₂₈H₃₂F₂N₂O·2HCl
Molecular Weight: 523,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00055193
CAS-Nummer: 67469-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR