(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride

Lieferant: Thermo Scientific
Gefahr

Synonyme: Ethambutol Hydrochloride, (2S,2'S)-2,2'-(Ethane-1,2-diylbis(azanediyl))bis(butan-1-ol) dihydrochloride, (S,S)-2,2'-(1,2-Ethanediyldiimino)bis-1-butanol Dihydrochloride

J60695.14
J60695.14EA 155 EUR
J60695.14
(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride
(S,S)-N,N'-Bis(1-hydroxy-2-butyl)ethylenediamine dihydrochloride

CAS No.: 1070-11-7 Molecular Formula: C10H24N2O2.2HCl Formula Weight: 277.23 Physical Form: Powder Appearance: White Solubility: Slightly soluble in water MDL No.: MFCD00216025


Applications: An antibacterial agent

Formel: C₁₀H₂₄N₂O₂·₂HCl
Molecular Weight: 277,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00216025
CAS-Nummer: 1070-11-7
EINECS: 213-970-7
Merck Index: 14,3720

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR