2'-Deoxyguanosine monohydrate

Lieferant: Thermo Fisher Scientific

J60741.03 J60741.06 J60741.09
J60741.03EA 68 EUR
J60741.03 J60741.06 J60741.09
2'-Deoxyguanosine monohydrate
2'-Deoxyguanosine monohydrate

CAS No.: 961-07-9 Molecular Formula: C10H13N5O4 Formula Weight: 267.25 Physical Form: Crystalline powder Appearance: White to off-white

Formel: C₁₀H₁₃N₅O₄·H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150760
CAS-Nummer: 312693-72-4
EINECS: 213-505-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR