Resveratrol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
J60790.ME
J60790.MEEA
403
EUR
J60790.ME
Resveratrol ≥98%
Resveratrol
Purity: 98%
CAS No.: 501-36-0
Molecular Formula: C14H12O3
Formula Weight: 228.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Merck Reference: 14,8158
MDL No.: MFCD00133799
A phytoestrogen with antitumor, antioxidant, antiplatelet, anti-inflammatory effects. Inhibits cytochrome P450 1A1 (IC50 = 23 uM) and displays mixed agonist/antagonist actions at ER-alpha and ER-beta estrogen receptors
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228.25 g/mol Schmelzpunkt: 260 Dichte: 1.36 g/cm³ (20 °C) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00133799 CAS-Nummer: 501-36-0 Merck Index: 12,08326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...