CGS 21680 hydrochloride 99%
Lieferant: Thermo Scientific
Synonyme:
3-(4-(2-((6-Amino-9-((2R,3R,4S,5S)-5-(ethylcarbamoyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-2-yl)amino)ethyl)phenyl)propanoic acid hydrochloride
J60793.MA
J60793.MAEA
329
EUR
J60793.MA
CGS 21680 hydrochloride 99%
CGS 21680 hydrochloride
Purity: 99%
CAS No.: 124182-57-6
Molecular Formula: C23H29N7O6.HCl
Formula Weight: 535.98
Physical Form: Solid
Appearance: White
MDL No.: MFCD11045878
Applications: An A2A Adenosine receptor agonist
Formel:
C₂₃H₂₉N₇O₆·HCl Molecular Weight: 535,98 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11045878 CAS-Nummer: 120225-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...