(-)-Epicatechin gallate

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3R)-2-(3,4-Dihydroxyphenyl)-5,7-dihydroxychroman-3-yl 3,4,5-trihydroxybenzoate, (-)-cis-3,3',4',5,7-Pentahydroxyflavane 3-Gallate

J60814.MA
J60814.MAEA 195 EUR
J60814.MA
(-)-Epicatechin gallate
(-)-Epicatechin gallate

CAS No.: 1257-08-5 Molecular Formula: C22H18O10 Formula Weight: 442.37 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00075936


Applications: A natural product from green tea that induces apoptosis

Formel: C₂₂H₁₈O₁₀
Molecular Weight: 442.38 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00075936
CAS-Nummer: 1257-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR