Buffer, potassium acetate, 1 M aq, soln, pH 7,5, RNAse free

Lieferant: Thermo Fisher Scientific

J60832.AK
J60832.AKEA 80.4 EUR
J60832.AK
Buffer, potassium acetate, 1 M aq, soln, pH 7,5, RNAse free
Puffer Klonpuffer

CAS No.: 127-08-2
Molecular formula: C₂H₃KO₂
Formula weight: 98,15
Physical form: liquid
Appearance: colourless

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR