Rifampicin für die Molekularbiologie
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5,6,9,17,19,21-Hexahydroxy-23-methoxy- 2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl- 1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca- [1,11,13]trienimino)-naphtho[2,1-b]furan- 1,11(2H)-dion-21-acetat, Rifampin
CAS No.: 13292-46-1
Molecular Formula: C43H58N4O12
Formula Weight: 822.94
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Faint orange to red/brown
Melting Point: 183-188°
Solubility: Soluble in DMSO or methanol
Beilstein Reference: 5723476
Merck Reference: 14,8216
MDL No.: MFCD00151389
Antibiotic effective against mycobacteria. Can also inhibit DNA polymerase
Formel:
C₄₃H₅₈N₄O₁₂ Schmelzpunkt: 183…188 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00151389 CAS-Nummer: 13292-46-1 EINECS: 236-312-0 Merck Index: 14,8216 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...