Dantrolene sodium salt

Lieferant: Thermo Scientific

Synonyme: Sodium 3-(((5-(4-nitrophenyl)furan-2-yl)methylene)amino)-2,5-dioxoimidazolidin-1-ide

J60887.MC
J60887.MCEA 70.5 EUR
J60887.MC
Dantrolene sodium salt
Dantrolene sodium salt

CAS No.: 14663-23-1 Molecular Formula: C14H9N4NaO5 Formula Weight: 336.23 Physical Form: Crystalline powder Appearance: Yellow to dark orange MDL No.: MFCD00079130


Applications: Inhibits intracellular calcium release from the sarcoplasmic reticulum

Formel: C₁₄H₉N₄NaO₅
Molecular Weight: 336,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00079130
CAS-Nummer: 14663-23-1
EINECS: 238-706-8
Merck Index: 14,2816

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR