Hexestrol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4,4'-(1,2-Diethylethylene)diphenol, rel-4,4'-((3R,4S)-Hexane-3,4-diyl)diphenol
J60889.03
J60889.03EA
53.9
EUR
J60889.03
Hexestrol ≥98%
Hexestrol
Purity: 98+%
CAS No.: 84-16-2
Molecular Formula: C18H22O2
Formula Weight: 270.37
Physical Form: Powder
Appearance: White
Melting Point: 186°
Beilstein Reference: 3209460
MDL No.: MFCD00068996
Applications: Nonsteroidal synthetic estrogen
Formel:
C₁₈H₂₂O₂ Molecular Weight: 270,37 g/mol Schmelzpunkt: 186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068996 CAS-Nummer: 84-16-2 EINECS: 201-518-1 Merck Index: 14,4701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...