Nω-Methyl-L-arginine monoacetate ≥99%

Lieferant: Thermo Scientific

Synonyme: H-Arg(Me)-OH.AcOH, N5-(Methylamidino)-L-ornithine acetate salt, Nomega-Monomethyl-L-arginine acetate, L-NMMA, Ng-Methyl-L-Arginine Acetate Salt, Nomega-Methyl-L-arginine acetate, N-ω-Monomethyl-L-arginine monoacetate, NG-Monomethyl-L-arginine acetate salt

J60890.MF
J60890.MFEA 298 EUR
J60890.MF
Nω-Methyl-L-arginine monoacetate ≥99%
Nω-Methyl-L-arginine monoacetate

Purity: 99+% CAS No.: 53308-83-1 Molecular Formula: C9H20N4O4 Formula Weight: 248.28 Physical Form: Powder Appearance: White to off-white Beilstein Reference: 6674255 MDL No.: MFCD00069311


Applications: Nitric oxide synthase inhibitor

Formel: C₇H₁₆N₄O₂·CH₃COOH
Molecular Weight: 248,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069311
CAS-Nummer: 53308-83-1
Merck Index: 14,6022

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR