Novobiocin sodium salt ≥95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Topoisomerase II inhibitor.
Novobiocin sodium salt is effective an antistaphylococcal agent used in the treatment of Methicillin-resistant Staphylococcus aureus (MRSA). It is also used to inhibit the activity of retroviral RNA-dependent DNA polymerase as well as inhibitor of bacterial DNA gyrase and eukaryotic DNA topoisomerase.
Applications: Topoisomerase II inhibitor
Formel:
C₃₁H₃₅N₂NaO₁₁ Schmelzpunkt: 215 °C |
MDL:
MFCD00066541 CAS-Nummer: 1476-53-5 EINECS: 216-023-6 Merck Index: 14,6722 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...