Zinksulfat 100 mM in wässriger Lösung
Lieferant: Thermo Scientific
|
Achtung
|
J60963.AE
J60963.AEEA
63.3
EUR
J60963.AE
Zinksulfat 100 mM in wässriger Lösung
Zinksulfat
CAS No.: 7733-02-0
Molecular Formula: O4SZn
Formula Weight: 161.43
Physical Form: Liquid
Appearance: Colourless
UN No.: 3082
MDL No.: MFCD00011302
Formel:
O₄SZn Molecular Weight: 161,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011302 CAS-Nummer: 7733-02-0 EINECS: 231-793-3 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...