Vinpocetine 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(41S,13aS)-Ethyl 13a-ethyl-2,3,41,5,6,13a-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate
J61000.MC
J61000.MCEA
378
EUR
J61000.MC
Vinpocetine 98%
Vinpocetine
Purity: 98%
CAS No.: 42971-09-5
Molecular Formula: C22H26N2O2
Formula Weight: 350.46
Physical Form: Powder
Appearance: White
Melting Point: 147-149°
MDL No.: MFCD00211233
Applications: Specific calcium-calmodulin-dependent phosphodiesterase inhibitor
Formel:
C₂₂H₂₆N₂O₂ Molecular Weight: 350,46 g/mol Schmelzpunkt: 147…149 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00211233 CAS-Nummer: 42971-09-5 EINECS: 256-028-0 Merck Index: 14,9991 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...