Octyl-β-D-thioglucopyranosid ≥98%
Lieferant: Thermo Fisher Scientific
J61028.06
J61028.09
J61028.88
J61028.06EA
295
EUR
J61028.06
J61028.09
J61028.88
Octyl-β-D-thioglucopyranosid ≥98%
Octyl-β-D-thioglucopyranosid
Purity: 98+%
CAS No.: 85618-21-9
Molecular Formula: C14H28O5S
Formula Weight: 308.44
Physical Form: Crystalline powder
Appearance: White to off-white
Melting Point: 128°
Solubility: Soluble in ethanol, water or methanol
Beilstein Reference: 4182783
MDL No.: MFCD00012189
Non-ionic detergent used for the solubilization of membrane proteins
Formel:
C₁₄H₂₈O₅S Schmelzpunkt: 128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012189 CAS-Nummer: 85618-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...