Castanospermine 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(1S,6S,7R,8R,8aR)-Octahydroindolizine-1,6,7,8-tetraol
J61071.MC
J61071.ME
J61071.MCEA
193
EUR
J61071.MC
J61071.ME
Castanospermine 99%
Castanospermine
Purity: 99%
CAS No.: 79831-76-8
Molecular Formula: C8H15NO4
Formula Weight: 189.21
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to off-white
Melting Point: 213-217°
Solubility: Soluble in water
Beilstein Reference: 3588654
MDL No.: MFCD00017555
Applications: Potent alpha and beta-glucosidase inhibitor.
Formel:
C₈H₁₅NO₄ Molecular Weight: 189.2 g/mol Schmelzpunkt: 213…217 °C |
MDL:
MFCD00017555 CAS-Nummer: 79831-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...