Jenner's stain Färbemittel für Hämatologie

Lieferant: Thermo Scientific

Synonyme: Eosin−Methylene Blue

J61077.14
J61077.14EA 23 EUR
J61077.14
Jenner's stain Färbemittel für Hämatologie
Jenner's stain

CAS No.: 62851-42-7 Physical Form: Powder Appearance: Dark green MDL No.: MFCD00081733


Applications: For use in staining blood smears

Lagertemperatur: Raumtemperatur
MDL: MFCD00081733
CAS-Nummer: 62851-42-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR