NaOH/SDS Lysis Lösung-Lösung, Ultrareine Qualität
Lieferant: VWR Chemicals
J611-500MLEA
373
EUR
J611-500ML
NaOH/SDS Lysis Lösung-Lösung, Ultrareine Qualität
NaOH/SDS Lysis
An ultra pure reagent for alkaline lysis of bacterial cells used in standard DNA purification procedures.
Lagertemperatur:
Raumtemperatur |
UN:
1824 ADR: 8 III |
Spezifikation Testergebnisse
Conductivity (10%, Water) @25 °C | Passes Test |
Titration | 7,6 – 8,4 g/l |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...