1-(3-Chlorphenyl)-2-[(2-methyl-2-propanyl)amino]-1-propanonhydrochlorid 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Amfebutamone hydrochloride, Bupropion Hydrochloride, Bupropion hydrochloride, 2-(tert-Butylamino)-1-(3-chlorophenyl)-1-propanone hydrochloride
J61105.03
J61105.03EA
187
EUR
J61105.03
1-(3-Chlorphenyl)-2-[(2-methyl-2-propanyl)amino]-1-propanonhydrochlorid 99%
2-(tert-Butylamino)-1-(3-chlorophenyl)propan-1-one hydrochloride
Purity: 99%
CAS No.: 31677-93-7
Molecular Formula: C13H18ClNO.HCl
Formula Weight: 276.20
Physical Form: Powder
Appearance: White
Melting Point: 233-234°
MDL No.: MFCD00055209
Applications: A selective inhibitor of dopamine uptake
Formel:
C₁₃H₁₉Cl₂NO Molecular Weight: 276,20 g/mol Schmelzpunkt: 233…234 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055209 CAS-Nummer: 31677-93-7 EINECS: 250-759-9 Merck Index: 14,1499 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...