Atenolol

Lieferant: Thermo Scientific

Synonyme: 2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide

J61199.03
J61199.03EA 67.3 EUR
J61199.03
Atenolol
Atenolol

CAS No.: 29122-68-7 Molecular Formula: C14H22N2O3 Formula Weight: 266.34 Physical Form: Powder Appearance: White Melting Point: 151-155° MDL No.: MFCD00057645


Applications: Cardioselective beta-adrenergic blocker. An antihypertensive

Formel: C₁₄H₂₂N₂O₃
Molecular Weight: 266,34 g/mol
Schmelzpunkt: 151…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00057645
CAS-Nummer: 29122-68-7
EINECS: 249-451-7
Merck Index: 14,859

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR