(+)-1-Deoxynojirimycin hydrochloride

Lieferant: Thermo Scientific

Synonyme: 1-Deoxymannojirimycin hydrochloride

J61277.MA
J61277.MAEA 239 EUR
J61277.MA
(+)-1-Deoxynojirimycin hydrochloride
(+)-1-Deoxynojirimycin hydrochloride

CAS No.: 73465-43-7 Molecular Formula: C6H13NO4.HCl Formula Weight: 199.60 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00083611


Applications: Selective inhibitor of a-mannosidase

Formel: C₆H₁₄ClNO₄
Molecular Weight: 199,64 g/mol
MDL: MFCD00083611
CAS-Nummer: 73465-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR