Amoxicillin trihydrate
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
6-[2-Amino-2-(4-hydroxyphenyl)-acetyl]amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, (2S,5R,6R)-6-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate, Amoxicillin Ttrihydrate, (2S,5R,6S)-6-((S)-2-amino-2-(4-hydroxyphenyl)acetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
CAS No.: 61336-70-7
Molecular Formula: C16H19N3O5S.3H2O
Formula Weight: 419.45 (365.40anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water at 3.4mg/ml.
Beilstein Reference: 7507120
MDL No.: MFCD00072029
Applications: A semi-synthetic antibiotic similar to penicillin
Formel:
C₁₆H₁₉N₃O₅S |
MDL:
MFCD00072029 CAS-Nummer: 61336-70-7 EINECS: 243-003-8 Merck Index: 14,577 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...