4-Aminobuttersäure ≥99%
Lieferant: Thermo Scientific
Synonyme:
γ-Aminobuttersäure, GABA
J61307.09
J61307.09EA
25.6
EUR
J61307.09
4-Aminobuttersäure ≥99%
4-Aminobuttersäure
Purity: 99+%
CAS No.: 56-12-2
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Physical Form: Powder
Appearance: White
Odour: Odourless
Melting Point: ca 195° dec.
Beilstein Reference: 906818
Merck Reference: 14,430
MDL No.: MFCD00008226
Formel:
C₄H₉NO₂ Molecular Weight: 103,12 g/mol Siedepunkt: 248 °C (1013 hPa) Schmelzpunkt: ∼ 195 °C (Zersetzung) Dichte: 1,21 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008226 CAS-Nummer: 56-12-2 EINECS: 200-258-6 Merck Index: 14,430 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...