Ciprofloxacin 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid

J61317.06 J61317.14
J61317.06EA 50.8 EUR
J61317.06 J61317.14
Ciprofloxacin 98%
Ciprofloxacin

Purity: 98% CAS No.: 85721-33-1 Molecular Formula: C17H18FN3O3 Formula Weight: 331.30 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white Melting Point: 253-257° Solubility: Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO Beilstein Reference: 3568352 MDL No.: MFCD00185755


Applications: An anti-bacterial agent effective against anaerobic bacteria

Formel: C₁₇H₁₈FN₃O₃
Molecular Weight: 331.35 g/mol
Schmelzpunkt: 253…257 °C
MDL: MFCD00185755
CAS-Nummer: 85721-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR