Ciprofloxacin 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
Purity: 98%
CAS No.: 85721-33-1
Molecular Formula: C17H18FN3O3
Formula Weight: 331.30
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Melting Point: 253-257°
Solubility: Soluble in 0.1N HCl at 25mg/ml. Poorly soluble in DMSO
Beilstein Reference: 3568352
MDL No.: MFCD00185755
Applications: An anti-bacterial agent effective against anaerobic bacteria
Formel:
C₁₇H₁₈FN₃O₃ Molecular Weight: 331.35 g/mol Schmelzpunkt: 253…257 °C |
MDL:
MFCD00185755 CAS-Nummer: 85721-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...