4-Fluor-7-nitrobenzofurazan
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F
J61336.MC
J61336.MCEA
447
EUR
J61336.MC
4-Fluor-7-nitrobenzofurazan
4-Fluor-7-nitrobenzofurazan
CAS No.: 29270-56-2
Molecular Formula: C6H2FN3O3
Formula Weight: 183.10
Flash Point: 110°(230°F)
Physical Form: Powder
Appearance: Yellow
Melting Point: 53-57°
Beilstein Reference: 1077571
MDL No.: MFCD00010196
Useful for amino acid and amine fluorescent labeling in HPLC
Formel:
C₆H₂FN₃O₃ Molecular Weight: 183,1 g/mol Schmelzpunkt: 53…57 °C Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010196 CAS-Nummer: 29270-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...