3-(4-Hydroxyphenyl)-7-chromanol

Lieferant: Thermo Scientific

Synonyme: 3-(4-Hydroxyphenyl)chroman-7-ol, (R,S)-Equol, 4′,7-Dihydroxyisoflavane, 7-Hydroxy-3-(4-hydroxyphenyl)chroman, (±)-Equol, 4',7-Isoflavandiol, 3-(4-Hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol

J61383.MC
J61383.MCEA 164 EUR
J61383.MC
3-(4-Hydroxyphenyl)-7-chromanol
(+/-)-Equol

CAS No.: 94105-90-5 Molecular Formula: C15H14O3 Formula Weight: 242.27 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Sensitivity: Hygroscopic Appearance: White to off-white Melting Point: 172-176° Beilstein Reference: 87752 MDL No.: MFCD00016662


Applications: Urinary metabolite of daidzein, produced by humans and horses

Formel: C₁₅H₁₄O₃
Molecular Weight: 242,27 g/mol
Schmelzpunkt: 172…176 °C
MDL: MFCD00016662
CAS-Nummer: 94105-90-5
Merck Index: 14,3644

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR