Alendronate sodium trihydrate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Amino-1-hydroxy-1-phosphonobutyl)phosphonic Acid Monosodium Salt, Sodium hydrogen (4-amino-1-hydroxy-1-phosphonobutyl)phosphonate trihydrate, Alendronic Acid Monosodium Salt, Alendronate sodium hydrate, Monosodium (4-Amino-1-hydroxy-1-phosphonobutyl)phosphonate
Purity: 97%
CAS No.: 121268-17-5
Molecular Formula: C4H12NaNO7P2.3H2O
Formula Weight: 325.12 (271.08anhy)
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White
Solubility: Soluble in water at 10mg/ml
MDL No.: MFCD01748233
Applications: Inhibits CD45 protein tyrosine phosphatase and farnesyl diphosphate synthase; induces apoptosis
Formel:
C₄H₁₂NaNO₇P₂·3H₂O Molecular Weight: 325,12 g/mol |
MDL:
MFCD01748233 CAS-Nummer: 121268-17-5 Merck Index: 14,229 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...