di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP) 5 mg, Tabletten
Lieferant: Thermo Scientific
Synonyme:
pNPP, ortho-Phosphorsäure(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, 4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, p-Nitrophenylphosphat Dinatriumsalz Hexahydrat, ortho-Phosphorsäuremono(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, Phosphatasesubstrat, 4-Nitrophenyldihydrogenphosphat Dinatriumsalz Hexahydrat
Substrate for alkaline and acid phosphatase determinations.
4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
Substrate for alkaline and acid phosphatase determinations
Formel:
C₆H₄NNa₂O₆P·6H₂O Molecular Weight: 371,144 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00066288 CAS-Nummer: 333338-18-4 EINECS: 224-246-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...