Tulobuterol hydrochloride 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-tert-Butylamino-1-(2-chlorophenyl)ethanol Hydrochloride

J61448.03
J61448.03EA 131 EUR
J61448.03
Tulobuterol hydrochloride 98%
Tulobuterol hydrochloride

Purity: 98% CAS No.: 56776-01-3 Molecular Formula: C12H18ClNO.HCl Formula Weight: 264.19 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 159-163° Solubility: Soluble in methanol or water. MDL No.: MFCD00214398


Applications: beta-2-adrenergic receptor agonist

Formel: C₁₂H₁₈ClNO
Molecular Weight: 264,19 g/mol
Schmelzpunkt: 159…163 °C
MDL: MFCD00214398
CAS-Nummer: 56776-01-3
Merck Index: 14,9811

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR