Methylgrün-Zinkchlorid-Doppelsalz
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J61509.14
J61509.22
J61509.14EA
101
EUR
J61509.14
J61509.22
Methylgrün-Zinkchlorid-Doppelsalz
Methylgrün-Zinkchlorid-Doppelsalz
CAS No.: 7114-03-6
Molecular Formula: C27H35Cl2N3.ZnCl2
Formula Weight: 653.24
Physical Form: Powder
Appearance: Dark green
Melting Point:
DNA stain useful as tracking dye in acid buffers for electrophoresis. Binds selectively to AT-rich regions of native DNA.
Formel:
C₂₇H₃₅BrClN₃·xZnCl₂ Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151094 CAS-Nummer: 7114-03-6 EINECS: 230-415-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...