Uranin

Lieferant: Thermo Fisher Scientific

Synonyme: 6-Hydroxy-9-(2-carboxyphenyl)-(3H)-xanthen-3-on Dinatriumsalz, 6-Hydroxy-9-(2-carboxyphenyl)- (3H)-xanthen-3-on Dinatriumsalz, Fluoreszein Dinatriumsalz, Fluorescein Natriumsalz, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)-benzoesäure Dinatriumsalz, Dinatrium-2-(3-oxo-6-oxidoxanthen-9-yl)benzoat, 6-Hydroxy-9-(2-carboxyphenyl)-(3H)-3-xanthenon Dinatriumsalz, Fluorescein-Natrium, Resorcinphthalein Dinatriumsalz, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoesäure Dinatriumsalz, Fluorescein Dinatriumsalz, 3′,6′-Dihydroxyspiro[2-benzofuran-3,9′-xanthen]-1-on Dinatriumsalz, 9-(2-Carboxyphenyl)-6-hydroxy-(3H)-3-xanthenon Dinatriumsalz

J61549.22 J61549.36
J61549.22EA 36.7 EUR
J61549.22 J61549.36
Uranin
Uranin

CAS No.: 518-47-8 Molecular Formula: C20H10Na2O5 Formula Weight: 376.28 Physical Form: Powder Appearance: Dark brownish-red to dark Solubility: Soluble in water at 1mg/ml Beilstein Reference: 3833041 Merck Reference: 14,4159 MDL No.: MFCD00167039


A fluorophore comonly used in microscopy

Formel: C₂₀H₁₀Na₂O₅
Schmelzpunkt: > 300 °C
Dichte: 1,601 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00167039
CAS-Nummer: 518-47-8
EINECS: 208-253-0
Merck Index: 14,4159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR