8-Cyclopentyl-1,3-dimethylxanthine

Lieferant: Thermo Fisher Scientific

J61565.MCEA 184 EUR
J61565.MC
8-Cyclopentyl-1,3-dimethylxanthine
8-Cyclopentyl-1,3-dimethylxanthine

CAS No.: 35873-49-5 Molecular Formula: C12H16N4O2 Formula Weight: 248.28 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 250-252° Solubility: Soluble in 0.1M NaOH. Slightly soluble in water. Also soluble in DMSO at 16mg/ml at 60°C. MDL No.: MFCD00055116


Applications: A selective A1 adenosine receptor antagonist

Formel: C₁₂H₁₆N₄O₂
Molecular Weight: 248.286 g/mol
Schmelzpunkt: 250…252 °C
MDL: MFCD00055116
CAS-Nummer: 35873-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR