Mianserin Hydrochlorid
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1,2,3,4,10,14b-Hexahydro-2-methyldibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride
J61570.MC
J61570.MCEA
90.5
EUR
J61570.MC
Mianserin Hydrochlorid
Mianserin Hydrochlorid
CAS No.: 21535-47-7
Molecular Formula: C18H20N2.HCl
Formula Weight: 300.83
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00055072
Formel:
C₁₈H₂₀N₂ Molecular Weight: 300,83 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055072 CAS-Nummer: 21535-47-7 EINECS: 244-426-7 Merck Index: 14,6172 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...