Spermidin Trihydrochlorid ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Azaoctamethylendiamin Trihydrochlorid, N-(3-Aminopropyl)-1,4-diaminobutan Trihydrochlorid, 4-Azoniaoctamethylendiammoniumtrichlorid
J61595.MC
J61595.MCEA
64
EUR
J61595.MC
Spermidin Trihydrochlorid ≥99%
Spermidin Trihydrochlorid
Purity: 99+%
CAS No.: 334-50-9
Molecular Formula: C7H19N3.3HCl
Formula Weight: 254.63
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 250° dec.
Solubility: Soluble in water
Beilstein Reference: 3552356
Merck Reference: 14,8742
MDL No.: MFCD00012918
Binds and precipitates DNA; may be used for purification of DNA binding proteins
Formel:
C₇H₂₂Cl₃N₃ Molecular Weight: 254,63 g/mol Schmelzpunkt: 250 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00012918 CAS-Nummer: 334-50-9 EINECS: 206-379-0 Merck Index: 14,8742 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...