Streptozocin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

J61601.ME J61601.03
J61601.MEEA 160 EUR
J61601.ME J61601.03
Streptozocin ≥97%
Streptozocin

Purity: 97+% CAS No.: 18883-66-4 Molecular Formula: C8H15N3O7 Formula Weight: 265.22 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to Off-white Melting Point: 121° Solubility: Soluble in water or 100% ethanol Beilstein Reference: 2060675 Merck Reference: 14,8832 MDL No.: MFCD00006607


Antibiotic and antitumor agent. Potent methylating agent for DNA

Formel: C₈H₁₅N₃O₇
Schmelzpunkt: 121 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00006607
CAS-Nummer: 18883-66-4
EINECS: 242-646-8
Merck Index: 14,8832

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR