Streptozocin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
J61601.ME
J61601.03
J61601.MEEA
160
EUR
J61601.ME
J61601.03
Streptozocin ≥97%
Streptozocin
Purity: 97+%
CAS No.: 18883-66-4
Molecular Formula: C8H15N3O7
Formula Weight: 265.22
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to Off-white
Melting Point: 121°
Solubility: Soluble in water or 100% ethanol
Beilstein Reference: 2060675
Merck Reference: 14,8832
MDL No.: MFCD00006607
Antibiotic and antitumor agent. Potent methylating agent for DNA
Formel:
C₈H₁₅N₃O₇ Schmelzpunkt: 121 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00006607 CAS-Nummer: 18883-66-4 EINECS: 242-646-8 Merck Index: 14,8832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...