Erbstatin analog

Lieferant: Thermo Scientific
Gefahr

Synonyme: Erbstatin Analog, 2,5-Dihydroxycinnamic acid methyll ester, Methyl 2,5-Dihydroxycinnamate, Methyl 3-(2,5-dihydroxyphenyl)acrylate

J61612.MA
J61612.MAEA 143 EUR
J61612.MA
Erbstatin analog
Erbstatin analog

CAS No.: 63177-57-1 Molecular Formula: C10H10O4 Formula Weight: 194.19 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Yellow Solubility: Soluble in methanol or 0.1M NaOH. Insoluble in water UN No.: UN2811 MDL No.: MFCD00132932


Applications: An inhibitor of the EGF receptor associated tyrosine kinase

Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
MDL: MFCD00132932
CAS-Nummer: 63177-57-1
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR