Adenosin-5'-monophosphat Dinatriumsalz (AMP Dinatriumsalz) Hydrat
Lieferant: Thermo Fisher Scientific
J61643.06
J61643.14
J61643.06EA
34.4
EUR
J61643.06
J61643.14
Adenosin-5'-monophosphat Dinatriumsalz (AMP Dinatriumsalz) Hydrat
Adenosin-5'-monophosphat Dinatriumsalz (AMP Dinatriumsalz) Hydrat
CAS No.: 4578-31-8
Molecular Formula: C10H12N5NaO7P
Formula Weight: 391.20
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 3586732
MDL No.: MFCD00065023
Formel:
C₁₀H₁₂N₅Na₂O₇P·H₂O Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065023 CAS-Nummer: 4578-31-8 EINECS: 224-961-2 Merck Index: 14,158 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...