Aniracetam
Lieferant: Thermo Fisher Scientific
Achtung
|
Synonyme:
1-(4-Methoxybenzoyl)pyrrolidin-2-one, 1-(4-Methoxybenzoyl)-2-pyrrolidone
J61661.03
J61661.03EA
98.8
EUR
J61661.03
Aniracetam
Aniracetam
CAS No.: 72432-10-1
Molecular Formula: C12H13NO3
Formula Weight: 219.24
Physical Form: Powder
Appearance: White to pale yellow
Solubility: Soluble in chloroform. Also soluble in ethanol. Insoluble in water
MDL No.: MFCD00153767
Applications: AMPA receptor potentiator. Slows the rate of ion-channel closing
Formel:
C₁₂H₁₃NO₃ Molecular Weight: 219.24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153767 CAS-Nummer: 72432-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...