13-cis-Retinoic acid
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 4759-48-2
Molecular Formula: C20H28O2
Formula Weight: 300.44
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Yellow-orange
Melting Point: 172-175°
MDL No.: MFCD00079542
Applications: A vitamin A analog that inhibits cell proliferation and induces cell differentiation
Formel:
C₂₀H₂₈O₂ Schmelzpunkt: 172…175 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00079542 CAS-Nummer: 4759-48-2 EINECS: 225-296-0 Merck Index: 14,5228 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...