Tyrphostin B46 ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)prop-2-enamide, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)-2-propenamide, AG 555, (E)-2-Cyano-3-(3,4-dihydroxyphenyl)-N-(3-phenylpropyl)acrylamide, Tyrphostin AG-555
J61704.MA
J61704.MAEA
134
EUR
J61704.MA
Tyrphostin B46 ≥98%
Tyrphostin B46
Purity: 98+%
CAS No.: 133550-34-2
Molecular Formula: C19H18N2O3
Formula Weight: 322.36
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Yellow
MDL No.: MFCD00209865
Applications: Inhibitor of EGF receptor kinase activity
Formel:
C₁₉H₁₈N₂O₃ Molecular Weight: 322.36 g/mol |
MDL:
MFCD00209865 CAS-Nummer: 133550-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...