XTT Natrium Salz

Lieferant: Thermo Scientific

Synonyme: 2,3-Bis(2-methoxy-4-nitro-5-sulphophenyl)-2H-tetrazolium-5-carboxanilide, XTT sodium, Sodium 5,5'-(5-(phenylcarbamoyl)-2H-tetrazole-3-ium-2,3-diyl)bis(4-methoxy-2-nitrobenzenesulphonate), sodium 2,3,-bis(2-methoxy-4-nitro-5-sulphophenyl)-5-[(phenylamino)-carbonyl]-2H-tetrazolium inner salt

J61726.MD
J61726.MDEA 365 EUR
J61726.MD
XTT Natrium Salz
XTT sodium salt

CAS No.: 111072-31-2 Molecular Formula: C22H16N7NaO13S2 Formula Weight: 673.52 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Yellow Solubility: Soluble in hot water at 2.5mg/ml MDL No.: MFCD00083517


Applications: Tetrazolium derivative useful in cancer research

Formel: C₂₂H₁₇N₇O₁₃S₂Na
Molecular Weight: 673,52 g/mol
MDL: MFCD00083517
CAS-Nummer: 111072-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR