Isoproterenol hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DL-Isoproterenol hydrochloride, (±)-Isoproterenol hydrochloride, Isoprenaline hydrochloride

J61788.14
J61788.14EA 208 EUR
J61788.14
Isoproterenol hydrochloride 98%
Isoproterenol hydrochloride

Purity: 98% CAS No.: 51-30-9 Molecular Formula: C11H17NO3.HCl Formula Weight: 247.72 Physical Form: Crystalline powder Appearance: White to yellow Melting Point: 165-175° dec. Beilstein Reference: 3917363 MDL No.: MFCD00012603


Applications: Standard selective beta-adrenoceptor agonist

Formel: C₁₁H₁₇NO₃
Molecular Weight: 247.72 g/mol
Schmelzpunkt: 165…175 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012603
CAS-Nummer: 51-30-9
EINECS: 200-089-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR