Natriumchlorid 5 M aqueous solution pH 8,0

Lieferant: Thermo Fisher Scientific

Synonyme: Saline, NaCl, Sea salt, Halite

J61807.AK
J61807.AKEA 55.9 EUR
J61807.AK
Natriumchlorid 5 M aqueous solution pH 8,0
Natriumchlorid

Sodium chloride suitable for use in cell culture applications.

Formel: NaCl
Molecular Weight: 58.44 g/mol
Siedepunkt: 1413
Schmelzpunkt: 801
Lagertemperatur: Raumtemperatur
MDL: MFCD00003477
CAS-Nummer: 7647-14-5
Merck Index: 14,08599

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR