Bafilamycin A1
Lieferant: Thermo Scientific
|
Achtung
|
J61835.MX
J61835.MXEA
364
EUR
J61835.MX
Bafilamycin A1
Bafilamycin A1
Bafilomycin A1 is a specific potent inhibitor of vacuolar ATPases. Used as an antibacterial, antifungal, antineoplastic, an immunosuppressive and also prevents maturation of autophagic vacuoles by inhibiting fusion between autophagosomes and lysosomes and is a macrolide antibiotic and potent and selective inhibitor of vacuolar-type H+ ATPase (V-ATPase).
- Solid or film
Soluble in methanol, ethanol, acetone and chloroform. Insoluble in water
Formel:
C₃₅H₅₈O₉ Lagertemperatur: Gefrierschrank |
MDL:
MFCD06795130 CAS-Nummer: 88899-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...