Phosphate, 0,2 M buffer solution, pH 7,6

Lieferant: Thermo Fisher Scientific

J61870.AP
J61870.APEA 39.9 EUR
J61870.AP
Phosphate, 0,2 M buffer solution, pH 7,6
Puffer Biologische Puffer

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR